Orders LinkContactUsLinkComponent
Stoppers, PTFE
Stoppers, PTFE
Bílá, 10/19
Katalogové číslo BOHLH936-02
Dodavatel:   Bohlender
Stoppers, PTFE
Katalogové číslo BOHLH936-02
Dodavatel:  Bohlender
Stoppers, PTFE
Stoppers, PTFE
Bílá, 10/19
Přejít na:
Price
201,00 Kč
Přihlásit se pro dostupnost
1 * 1 KS
Product Details & Documents
For closing ground joint parts. With ground joint and sealing rings on the outside; knurled or hexagonal grip. Compared with glass stoppers, they are easily removable and can be used without grease.
  • PTFE
  • Temperature resistance from −200 to + 250 °C
  • Chemical resistance
  • FDA conform
The stoppers can expand under heat which might lead to a breaking of the ground joint sockets.
Specifikace
  • NZ:
    10/19
  • Barva:
    Bílá