Orders LinkContactUsLinkComponent
Telescopic handles
Telescopic handles
02, Násada hliníková teleskopická, 1000 - 1800 mm
Catalog # 129-0456
Supplier:   FHP VILEDA SCS
Telescopic handles
Catalog # 129-0456
Supplier:  FHP VILEDA SCS
Telescopic handles
Telescopic handles
02, Násada hliníková teleskopická, 1000 - 1800 mm
Price
698,00 Kč
Log in for availability
1 * 1 KS
Product Details & Documents
These lightweight, strong extendable handles are perfect for all controlled environments. Fully autoclavable.
  • Extremely easy to use and extend with a simple twist of the wrist
  • Resist autoclave, gamma and ETO processes
  • Ergonomic center and anti-slip grips
  • Available with colour coding clips (Blue, Red, Yellow, Green)
For use with all Vileda Professional frames and CE Squeegee.
Specifications
  • Popis:
    Násada hliníková teleskopická
  • Délka:
    1000 - 1800 mm
  • Sekvence:
    02