Orders LinkContactUsLinkComponent
Write-on labels
Write-on labels
20, Write-on labels, Vinyl, Černá na bílé, 150×100 mm, 10 labels/pack
Catalog # BRDY230632
Supplier:   Brady
Write-on labels
Catalog # BRDY230632
Supplier:  Brady
Write-on labels
Write-on labels
20, Write-on labels, Vinyl, Černá na bílé, 150×100 mm, 10 labels/pack
Price
863,00 Kč
Log in for availability
1 * 10 KS
Product Details & Documents
These write-on labels provide easy identification for calibration, testing, and repair of tools and equipment.
  • Labels are used to track calibration, inspection, or parts and/or equipement
Write-on areas allow date, inspector, or other information to be periodically added.
Specifications
  • Barva:
    Černá na bílé
  • Popis:
    Write-on labels
  • Rozměry:
    150×100 mm
  • Materiál:
    Vinyl
  • Sekvence:
    20
  • Balené:
    10 labels/pack