HPLC columns
05, Analytical, 2,10 mm, 250 mm
Catalog # JOURJR-65006NF
Supplier: VICI JOUR RESEARCH
Jump to:
Product Details & Documents
These PEEK columns are designed for naro-bore applications with small amount of samples.
- Biocompatible
- High pressure applications
- Packing devices available
Specifications
- Formát kolony:Analytical
- Ø vnitř.:2,10 mm
- Délka:250 mm
- Sekvence:05
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



