Sample lance
Valve lance, Stainless steel V4A, 800 mm, 32 mm
Catalog # 300-0077
Supplier: BURKLE
Jump to:
Product Details & Documents
Samplers for viscous, paste-like and semi-liquid substances.
- Optimised opening mechanism (can be completely disassembled)
- Easy to use and to clean
- Different layers remain intact particularly with paste-like substances
Designed for use in pharmaceutical applications.
: DIN 51750.3 and 53242.3
Specifications
- Ø:32 mm
- Popis:Valve lance
- Délka:800 mm
- Materiál:Stainless steel V4A