Orders LinkContactUsLinkComponent
Conductivity standards, KCL
02, Standardní roztok chloridu draselného, 0,01 M, 1,413 mS/cm, 500 mL
Catalog # HACHC20C270
Supplier:   Hach
Conductivity standards, KCL
Catalog # HACHC20C270
Supplier:  Hach
CAS Number:  
Conductivity standards, KCL
Conductivity standards, KCL
02, Standardní roztok chloridu draselného, 0,01 M, 1,413 mS/cm, 500 mL
CAS Number:  
Price
1 660,00 Kč
Log in for availability
1 * 500 mL
Product Details & Documents
The potassium chloride standard solutions are used for calibrating conductivity cells.
Specifications
Specifications
  • Bal.:
    500 mL
  • Popis:
    Standardní roztok chloridu draselného, 0,01 M
  • Sekvence:
    02
  • Vodivost:
    1,413 mS/cm
Specifications