Orders LinkContactUsLinkComponent
Rotary evaporator receiving flasks
Rotary evaporator receiving flasks
08, 500 ml, KS 35/20
Catalog # 531-0827
Supplier:   Lenz Laborglas
Rotary evaporator receiving flasks
Catalog # 531-0827
Supplier:  Lenz Laborglas
Rotary evaporator receiving flasks
Rotary evaporator receiving flasks
08, 500 ml, KS 35/20
Price
3 490,00 Kč
Log in for availability
1 * 1 KS
Product Details & Documents
DURAN®, borosilicate glass, clear or plastic-coated.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
Spherical joint KS 35/20, centred, suitable for all rotary evaporators.
Documents
Specifications
  • Zásuvka:
    KS 35/20
  • Kapacita:
    500 ml
  • Sekvence:
    08