Rectangular cover glasses
004, 22×32 mm, 0,16 - 0,19 mm
Katalogové číslo HECH92099005095
Dodavatel: GLASWARENFABRIK KARL HECHT
Přejít na:
Product Details & Documents
Cover glass D 263® M made of borosilicate glass.
- Free from colour distortion and highly transparent
- Excellent quality, low levels of inclusions, striae, bubbles and streaks
- High spectral transmission, excellent flatness, optimum resistance to chemicals
Certifications: ISO 8255
Specifikace
- D׊:22×32 mm
- Sekvence:004
- Tloušťka:0,16 - 0,19 mm