Orders LinkContactUsLinkComponent
Electroporation solution kits, BTXpress™
Electroporation solution kits, BTXpress™
15, BTXpress™ solution: 10 ml bottle for up to 100 reactions (without cuvettes), 10 mL
Katalogové číslo 732-1285
Dodavatel:   BTX
Electroporation solution kits, BTXpress™
Katalogové číslo 732-1285
Dodavatel:  BTX
Číslo CAS:  
Electroporation solution kits, BTXpress™
Electroporation solution kits, BTXpress™
15, BTXpress™ solution: 10 ml bottle for up to 100 reactions (without cuvettes), 10 mL
Přejít na:
Číslo CAS:  
Price
17 210,00 Kč
Přihlásit se pro dostupnost
1 * 10 mL
Product Details & Documents
The BTXpress™ is a single buffer solution, developed to quickly and efficiently deliver genes into mammalian cells that were previously considered hard to transfect by chemical and other non viral methods. Transfection using this high performance electroporation solution is equally effective in delivering DNA as well as siRNA into mammalian cells.
  • High efficiency transfection of cell lines and previously considered hard to transfect cells
  • Low toxicity resulting in improved cell viability
  • One buffer used in place of standard electroporation buffers for all mammalian cell types
  • Versatility of choosing electroporation optimisation settings for the highest transfection efficiency with BTX instruments
  • Increased numbers of tranfections per kit providing better value
As a universal solution the BTXpress™ electroporation reagent can be used with electroporation generators, including Amaxa™ Nucleofector®.
Informace o dodávce: BTXpress™ high performance electroporation solution is supplied either as a kit, which includes the BTX Plus cuvettes with transfer pipettes, or as a buffer alone.
Documents
Specifikace
Specifikace
  • Bal.:
    10 mL
  • Popis:
    BTXpress™ solution: 10 ml bottle for up to 100 reactions (without cuvettes)
  • Sekvence:
    15
Specifikace