BUFFER SOLUTION PH10 TWIN NECK 1 * 500 mL
REGE11005CTT
: REAGECON
Frequently Bought Together
Aceton ≥99%, TECHNICAL
: VWR Chemicals
VWR®, Cover Glasses, Square
: VWR Collection
Roztoky standardní pro měření vodivosti
: VWR Chemicals
VWR® AVS TITRINORM, pufrovací roztoky, 20 °C
: VWR Chemicals
BUFFER PH 4 ROOD 20-25 C 1 * 500 mL
: REAGECON
VWR®, Nitrile Gloves
: VWR Collection
VWR®, Screw Neck Vials ND24
: VWR Collection