Uzvr k vialce PP ND18 roub.ern,bez otvoru,Silikon modr/PTFE bl (100ks) 1 * 100 KS
Katalogové číslo LAPH18151132
Dodavatel: LA PHA PACK
Uzvr k vialce PP ND18 roub.ern,bez otvoru,Silikon modr/PTFE bl (100ks) 1 * 100 KS
Katalogové číslo LAPH18151132
Dodavatel: LA PHA PACK
Frequently Bought Together
Aceton ≥99%, TECHNICAL
Dodavatel: VWR Chemicals
Sealing film for manual application, Parafilm® M
Dodavatel: HEATHROW
Voda, HiPerSolv CHROMANORM® for HPLC
Dodavatel: VWR Chemicals
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Dodavatel: VWR Chemicals
VWR® NITRILE LIGHT, Rukavice, nitrilové
Dodavatel: VWR Collection
VWR®, Pipety Pasteurovy, High performance
Dodavatel: VWR Collection
Rukavice, nitrilové, Kimtech™ Purple Nitrile™
Dodavatel: KIMBERLY CLARK
VWR®, Sérologická pipeta, standardní
Dodavatel: VWR Collection
VWR®, Ubrousky čisticí
Dodavatel: VWR Collection
VWR®, Nitrile Gloves
Dodavatel: VWR Collection
Acetonitril ≥99.9%, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Dodavatel: VWR Chemicals