Uzávěr PP šroubovací ND9 otvor 6mm,Sil/PTFE proříz. (100ks) 1 * 100 KS
Katalogové číslo LAPH09150869
Dodavatel: LA PHA PACK
Uzávěr PP šroubovací ND9 otvor 6mm,Sil/PTFE proříz. (100ks) 1 * 100 KS
Katalogové číslo LAPH09150869
Dodavatel: LA PHA PACK
Frequently Bought Together
VWR®, Screw Caps for Short Thread Vials ND9
Dodavatel: VWR Collection
Voda, HiPerSolv CHROMANORM® for HPLC
Dodavatel: VWR Chemicals
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Dodavatel: VWR Chemicals
Aceton ≥99%, TECHNICAL
Dodavatel: VWR Chemicals
Acetonitril ≥99.9%, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Dodavatel: VWR Chemicals
VWR®, Nitrile Gloves
Dodavatel: VWR Collection
Voda, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Dodavatel: VWR Chemicals
2-Propanol ≥99.8%, HiPerSolv CHROMANORM® for HPLC
Dodavatel: VWR Chemicals
VWR® NITRILE LIGHT, Rukavice, nitrilové
Dodavatel: VWR Collection
Aceton ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Dodavatel: VWR Chemicals
Safety Bottle Container
Dodavatel: BOCHEM