[EN]ELECTROLYTE KNO3-1MOL/L 1 * 250 mL
Catalog # METR6.2310.010
Supplier: METROHM
Frequently Bought Together
Aceton ≥99%, TECHNICAL
Supplier: VWR Chemicals
Nitric acid 69% 69%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
VWR®, Nitrile Gloves
Supplier: VWR Collection
Jodid draselný ≥99.5%, AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
VWR®, Ubrousky čisticí
Supplier: VWR Collection
Chlorid draselný 3 mol/l (3 N) in aqueous solution, Supelco®
Supplier: Merck
VWR®, Pipety Pasteurovy, High performance
Supplier: VWR Collection
VWR® NITRILE LIGHT, Rukavice, nitrilové
Supplier: VWR Collection
VWR®, Snap Ring Vials ND11
Supplier: VWR Collection
Voda, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
Glass microfibre filters without binder, grade GF/C, Whatman™
Supplier: Whatman products (Cytiva)
[EN]ELECTROLYTE 3 MOL / L KCL 1 * 50 mL
Supplier: METROHM