Orders LinkContactUsLinkComponent
Autoloop Pro
Autoloop Pro
02, Loop sterilising carousel, 1200 g, Fuego series gas burners
Katalogové číslo WART8.000.400
Dodavatel:   WARTEWIG
Autoloop Pro
Katalogové číslo WART8.000.400
Dodavatel:  WARTEWIG
Autoloop Pro
Autoloop Pro
02, Loop sterilising carousel, 1200 g, Fuego series gas burners
Přejít na:
Price
32 450,00 Kč
Přihlásit se pro dostupnost
1 * 1 KS
Product Details & Documents
The inoculation loop carrousel Autoloop Pro, which is suitable for up to four inoculation loops, was developed to comply with current requirements in microbiological, cellbiological and biotechnological laboratories.
  • Stable housing enables comfortable and easy access to inoculation loops
  • Reduce gas consumption
  • Intelligent sensor of the Autoloop PRO monitors safe sterilisation
  • Automatic unit switch off after 1 hour
The Autoloop PRO rotates and controls flaming automatically, No unintentional use of hot inoculation loops: When flaming is completed, the display shows the remaining cooling time and the removal positions of cool inoculation loops. All the suspended inoculation loops can be flamed as needed in one full rotation of the carrousel.
 : Supplied without Fuego SCS series laboratory gas burner.
Specifikace
  • Popis:
    Loop sterilising carousel, 1200 g
  • Sekvence:
    02
  • Určeno pro:
    Fuego series gas burners