Orders LinkContactUsLinkComponent
Adapters, PTFE, socket on cone
Adapters, PTFE, socket on cone
11, , střední délka, B, 45/40 NS, 24/29 NS, ,
Catalog # COWI021.2445
Supplier:   COWIE
Adapters, PTFE, socket on cone
Catalog # COWI021.2445
Supplier:  COWIE
Adapters, PTFE, socket on cone
Adapters, PTFE, socket on cone
11, , střední délka, B, 45/40 NS, 24/29 NS, ,
Price
951,00 Kč
Log in for availability
1 * 1 KS
Product Details & Documents
Made of pure PTFE, white.
  • Very good chemical resistance
  • Very good temperature resistance
  • Fully adjustable with replacement Viton® seal
  • Contact materials are glass and PTFE only
This holder enables standard probes and thermometers to be assembled in standard taper laboratory glassware.
Specifications
  • Rozměry zábrusu:
    45/40 NS
  • Velikost objímky:
    24/29 NS
  • Type:
    střední délka, B
  • Sekvence:
    11