Orders LinkContactUsLinkComponent
Lock boards
Lock boards
02, Lockout station for 10 locks, Žlutá, 342×342 mm
Katalogové číslo 238-0192
Dodavatel:   Brady
Lock boards
Katalogové číslo 238-0192
Dodavatel:  Brady
Lock boards
Lock boards
02, Lockout station for 10 locks, Žlutá, 342×342 mm
Přejít na:
Price
8 670,00 Kč
Přihlásit se pro dostupnost
1 * 1 KS
Product Details & Documents
These acrylic lockout stations are suitable for storing tags, padlocks and hasps for either 5, 10, 26 or 36 people.
  • Highly visible thanks to yellow colour
  • For wall mounting
Please note: Tags, padlocks and hasps are not included.
Specifikace
  • Type:
    Lockout station for 10 locks
  • Barva:
    Žlutá
  • Sekvence:
    02
  • Š×H×V:
    342×342 mm