Orders LinkContactUsLinkComponent
Conical separating funnels, DURAN®
Conical separating funnels, DURAN®
100 ml, 19/26 NS
Catalog # SCOT1064805
Supplier:   DWK Life Sciences
Conical separating funnels, DURAN®
Catalog # SCOT1064805
Supplier:  DWK Life Sciences
Conical separating funnels, DURAN®
Conical separating funnels, DURAN®
100 ml, 19/26 NS
Price
2 210,00 Kč
Log in for availability
1 * 1 KS
Product Details & Documents
DURAN®, borosilicate glass 3.3, clear, with NS-PE stopper.
  • Very good chemical resistance
  • Very good temperature resistance
  • Wwithout graduation complies to DIN ISO 4800
With socket and plain stem. Available with interchangeable solid glass key or alternatively PTFE key for grease-free performance.
Specifications
  • Velikost objímky:
    19/26 NS
  • Kapacita:
    100 ml