2-(P-TOLUIDINO)NAPHTHALENE-6-SULFONIC*AC 1 * 250 mg
Katalogové číslo SIALT9792-250MG
Dodavatel: Merck
2-(P-TOLUIDINO)NAPHTHALENE-6-SULFONIC*AC 1 * 250 mg
Katalogové číslo SIALT9792-250MG
Dodavatel: Merck
Frequently Bought Together
Aceton ≥99.5%, GPR RECTAPUR®
Dodavatel: VWR Chemicals
Isooktán ≥99.5%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Dodavatel: VWR Chemicals
Methanol ≥99.9% (by GC), HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Dodavatel: VWR Chemicals
Tetrahydrofuran, LiChrosolv® for liquid chromatography, Supelco®
Dodavatel: Merck
Diethylether ≥99.7% stabilised, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Dodavatel: VWR Chemicals
Neutralising agent for the automated cleaning of glassware, neodisher® Z
Dodavatel: Dr. Weigert
Aceton ≥99.8%, HiPerSolv CHROMANORM® for HPLC
Dodavatel: VWR Chemicals
Methanol ≥99.8%, HiPerSolv CHROMANORM®, Isocratic Grade for HPLC
Dodavatel: VWR Chemicals
n-Hexan ≥95%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Dodavatel: VWR Chemicals
n-Hexan, SupraSolv® for GC-ECD/FID, Supelco®
Dodavatel: Merck