[EN]DIOCTYL SULFOSUCCINATE SODIUMSIGMAUL 1 * 100 g
Katalogové číslo SIALD4422-100G
Dodavatel: Merck
Frequently Bought Together
Ethanol 96% vol Ph. Eur.
Dodavatel: VWR Chemicals
Anilin, EMSURE® p.a., Supelco®
Dodavatel: Merck
1-Butanol ≥98.5%, GPR RECTAPUR®
Dodavatel: VWR Chemicals
Aceton, EMSURE® ACS, ISO, Reag. Ph. Eur. p.a., Supelco®
Dodavatel: Merck
2-Propanol ≥99.7%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Dodavatel: VWR Chemicals
2-Propanol ≥98%, TECHNICAL
Dodavatel: VWR Chemicals
Acetonitrile : water : trifluoroacetic acid (50 : 47,5 : 2,5), HiPerSolv CHROMANORM®
Dodavatel: VWR Chemicals
Dichlormethan, dehydrated (max. 0.01% H₂O) ≥99% stabilised, GPR RECTAPUR® for synthesis
Dodavatel: VWR Chemicals
Glycerin ≥98%, GPR RECTAPUR®
Dodavatel: VWR Chemicals
Isooktán, SupraSolv® for GC-ECD/FID, Supelco®
Dodavatel: Merck
2-Propanol ≥99.0%, GPR RECTAPUR®
Dodavatel: VWR Chemicals
Voda USP, JP, Ph. Eur., purified
Dodavatel: VWR Chemicals
Benzonitril 99+% for spectroscopy
Dodavatel: Thermo Fisher Scientific