2-Bromophenol, Sigma-Aldrich®
3368, Glass bottle for solids, 50 g
SIAL130915-50G
: Merck
- :95-56-7
- :C₆H₅BrO
- :MFCD00002146
- :173.01
- :13,01413
- :195 °C (1013 hPa)
- :5,6 °C
- :1,49 g/cm³ (20 °C)
- :Ambient
- :202-432-7
- :3082
- :9,III
- :M-Clarity™ quality level = MQ100
- :Density: 1,49 g/cm³ (20 °C)
- Typ balení:Glass bottle for solids
- Bal.:50 g
- Sekvence:3368
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.