2-BROMOETHANESULFONIC ACID, SODIUM SALT, 1 * 100 g
Katalogové číslo SIAL137502-100G
Dodavatel: Merck
2-BROMOETHANESULFONIC ACID, SODIUM SALT, 1 * 100 g
Katalogové číslo SIAL137502-100G
Dodavatel: Merck
Frequently Bought Together
VWR®, Nitrile Gloves
Dodavatel: VWR Collection
Acetonitril ≥99.9%, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Dodavatel: VWR Chemicals
Dihydrogenfosforečnan draselný ≥99%, powder, GPR RECTAPUR®
Dodavatel: VWR Chemicals
Detergents, liquid, for automated cleaning of glassware, neodisher® FLA
Dodavatel: Dr. Weigert
Aceton ≥99%, TECHNICAL
Dodavatel: VWR Chemicals
Voda, HiPerSolv CHROMANORM® for HPLC
Dodavatel: VWR Chemicals
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Dodavatel: VWR Chemicals
Glycerin ≥99.5%, AnalaR NORMAPUR® ACS analytical reagent, redistilled
Dodavatel: VWR Chemicals
2-Propanol ≥99.8%, HiPerSolv CHROMANORM® for HPLC
Dodavatel: VWR Chemicals
Chlorid sodný, EMSURE® ACS, ISO, Reag. Ph. Eur. p.a., Supelco®
Dodavatel: Merck
VWR®, Pipety Pasteurovy, High performance
Dodavatel: VWR Collection
Ethanol absolute ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Dodavatel: VWR Chemicals
Voda, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Dodavatel: VWR Chemicals