Orders LinkContactUsLinkComponent
Posts for shelving systems, solid, Super Erecta® SiteSelect™
Posts for shelving systems, solid, Super Erecta® SiteSelect™
37, Stationary post, SiteSelect™, chrome plated, 2200 mm height, Drop Mat™ Super Erecta® display shelving, Super Adjustable Super Erecta® shelving, Super Erecta® shelving, Super Erecta® Solid shelving, APPEAL™ display shelving
Katalogové číslo 139-0255
Dodavatel:   Metro International
Posts for shelving systems, solid, Super Erecta® SiteSelect™
Katalogové číslo 139-0255
Dodavatel:  Metro International
Posts for shelving systems, solid, Super Erecta® SiteSelect™
Posts for shelving systems, solid, Super Erecta® SiteSelect™
37, Stationary post, SiteSelect™, chrome plated, 2200 mm height, Drop Mat™ Super Erecta® display shelving, Super Adjustable Super Erecta® shelving, Super Erecta® shelving, Super Erecta® Solid shelving, APPEAL™ display shelving
Přejít na:
Price
1 520,00 Kč
Přihlásit se pro dostupnost
1 * 1 KS
Product Details & Documents
Specifikace
  • Popis:
    Stationary post, SiteSelect™, chrome plated, 2200 mm height
  • Sekvence:
    37
  • Určeno pro:
    Drop Mat™ Super Erecta® display shelving, Super Adjustable Super Erecta® shelving, Super Erecta® shelving, Super Erecta® Solid shelving, APPEAL™ display shelving