Robotic tips for Tecan®
907-021, 80 µl, +, +, Transparent, 10 racks, 96 each
732-1535
High quality tips, especially intended for research in molecular biology laboratories.
- Transparent or black PP
- Tips with ART® filters: Eliminate any risk of cross-contamination between samples; designed for applications with DNA, RNA as well as radioactive and infectious samples
- Certified free of RNase, DNase, DNA and pyrogens
- Sterile tips: Sterilised by gamma radiation
- Wide aperture tips: Designed for applications with more viscous samples
: Available in boxes of 10 racks or 50 racks (Econopak).
- Balené:10 racks, 96 each
- Sterilní:+
- Objem:80 µl
- Druh špičky:907-021
- Barva špičky:Transparent
- Filtr:+
- Cat. No.:732-1535
- Certifikát ochrany životního prostředí:IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
- Environmentally Preferable:Low Manufacturing Impact