Orders LinkContactUsLinkComponent
Percoll™
Percoll™
03, Percoll™, 6×1 l
Katalogové číslo 17-0891-09
Dodavatel:   Cytiva
Percoll™
Katalogové číslo 17-0891-09
Dodavatel:  Cytiva
Percoll™
Percoll™
03, Percoll™, 6×1 l
Přejít na:
Price
84 480,00 Kč
Přihlásit se pro dostupnost
1 * 6 KS
Product Details & Documents
Percoll® is commonly used for separating cells, organelles, viruses, and other subcellular particles.
  • For Research Use Only
  • Commonly used in basic research applications
  • Used in balanced salt solutions
  • Colorless to slightly yellow
A silica-based density gradient media for use in separating cells, organelles, viruses, and other subcellular particles. Percoll® comprises silica particles coated with polyvinylpyrrolidone (PVP) and is commonly used in basic research applications. Also used in balanced salt solutions, physiological saline or 0,25 M sucrose, and for formation of gradients either by the use of convenient gradient mixers or by high speed centrifugation. Colloidal solution of silica coated with polyvinylpyrrolidone (PVP). Liquid/suspension. Colorless to slightly yellow. Certificate of Analysis is available.
Upozornení: Percoll is for Research use only.
Specifikace
  • Popis:
    Percoll™, 6×1 l
  • Sekvence:
    03