Orders LinkContactUsLinkComponent
Adapters, reduction, Quickfit®
Adapters, reduction, Quickfit®
19/26 NS
Katalogové číslo 201-0112
Dodavatel:   SciLabware
Adapters, reduction, Quickfit®
Katalogové číslo 201-0112
Dodavatel:  SciLabware
Adapters, reduction, Quickfit®
Adapters, reduction, Quickfit®
19/26 NS
Přejít na:
Price
2 000,00 Kč
Přihlásit se pro dostupnost
1 * 2 KS
Product Details & Documents
PYREX®, borosilicate glass, Quickfit® joints, clear.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
Reduction adapter from large cone at the base to small socket on the top.
Specifikace
  • Velikost objímky:
    19/26 NS