Orders LinkContactUsLinkComponent
Forceps, precision
Forceps, precision
02, Precision forceps, extra sharp, curved, Curved, 105 mm
Katalogové číslo BOCH1940
Dodavatel:   BOCHEM
Forceps, precision
Katalogové číslo BOCH1940
Dodavatel:  BOCHEM
Forceps, precision
Forceps, precision
02, Precision forceps, extra sharp, curved, Curved, 105 mm
Přejít na:
Price
1 550,00 Kč
Přihlásit se pro dostupnost
1 * 1 KS
Product Details & Documents
These non serrated forceps are made of durable titanium.
  • High corrosion and thermal resistance
  • Lightweight, high strength
Titanium is resistant to diluted sulphuric acid, hydrochloric acid, solutions containing chloride, cold nitric acid as well as most organic acids and alkaline solutions, such as sodium hydroxide.
Specifikace
  • Type:
    Curved
  • Popis:
    Precision forceps, extra sharp, curved
  • Délka:
    105 mm
  • Sekvence:
    02