Orders LinkContactUsLinkComponent
Support clips
Support clips
Support clip with fitting, O. Ø 133 - 140 mm
Katalogové číslo BOCH600
Dodavatel:   BOCHEM
Support clips
Katalogové číslo BOCH600
Dodavatel:  BOCHEM
Support clips
Support clips
Support clip with fitting, O. Ø 133 - 140 mm
Přejít na:
Price
3 550,00 Kč
Přihlásit se pro dostupnost
1 * 1 KS
Product Details & Documents
These support clips are coated with ceramic inside.
  • For tubes and cylindrical vessels
  • With fitting
Informace pro objednávání: The shafts (rods) need to be ordered separately.
Specifikace
  • Popis:
    Support clip with fitting, O. Ø 133 - 140 mm