Support clips
Support clip with fitting, O. Ø 58 - 61 mm
Katalogové číslo BOCH594
Dodavatel: BOCHEM
Přejít na:
Product Details & Documents
These support clips are coated with ceramic inside.
- For tubes and cylindrical vessels
- With fitting
Informace pro objednávání: The shafts (rods) need to be ordered separately.
Specifikace
- Popis:Support clip with fitting, O. Ø 58 - 61 mm