Orders LinkContactUsLinkComponent
Watch glasses
Watch glasses
17, Watch glass, glass, with fused rim, 110 mm
Katalogové číslo HECH40010011
Watch glasses
Katalogové číslo HECH40010011
Watch glasses
Watch glasses
17, Watch glass, glass, with fused rim, 110 mm
Přejít na:
Price
589,00 Kč
Přihlásit se pro dostupnost
1 * 10 KS
Product Details & Documents
Evaporating dishes, made of glass, with fused rim.
  • Good chemical restistance
  • Available with or without hole
Certifications: DIN 12 341
Specifikace
  • Ø:
    110 mm
  • Popis:
    Watch glass, glass, with fused rim
  • Sekvence:
    17