Orders LinkContactUsLinkComponent
Safety spectacles, Tracker
Safety spectacles, Tracker
01, Anti-UV, anti-fog, anti-scratch, Černá, Smoke PC
Katalogové číslo 111-0461
Dodavatel:   Bollé Safety
Safety spectacles, Tracker
Katalogové číslo 111-0461
Dodavatel:  Bollé Safety
Safety spectacles, Tracker
Safety spectacles, Tracker
01, Anti-UV, anti-fog, anti-scratch, Černá, Smoke PC
Přejít na:
Price
502,00 Kč
Přihlásit se pro dostupnost
1 * 1 KS
Product Details & Documents
These sporty design safety spectacles feature a durable PC frame, a smoke PC dual lens, straight temples with upper and lower protection and an adjustable headband.
  • Interchangeable foam reinforcement with indirect ventilation
  • Adjustable head strap and rubber arm tips provide a secure fit and greater protection
  • ALS anti-fog/anti-scratch coating on both sides of clear and smoke lenses
  • Rubberised eye seal provides exceptional protection against dust and other particles
Weight: 52 g
EN 166
Certifications: EN 166
Delivery information: Supplied with microfibre pouch.
Specifikace
  • Type:
    Anti-UV, anti-fog, anti-scratch
  • Barva:
    Černá
  • Sekvence:
    01
  • Čočky/objektiv:
    Smoke PC