Spin Adapters for Gravity Columns
06, PD-10 Spin adapter, for PD-10 desalting columns
Katalogové číslo 28-9232-45
Dodavatel: Cytiva
Přejít na:
Product Details & Documents
Convenient and rapid cleanup with high reproducibility and the potential for high throughput by running multiple samples in parallel, High desalting capacity, The use of centrifugation protocol with all gravity columns.
- Convenient adapters, enable spinning of all gravity columns
Adapters are polypropylene rings designed for use with the gravity columns PD MiniTrap G-25, PD MidiTrap G-25, and PD-10 Desalting Columns when the spin protocol is used. MiniSpin Adapter fits PD MiniTrap G-25 columns, MidiSpin Adapter fits PD MidiTrap G-25 columns, and PD-10 Spin Adapter fits PD-10 Desalting Columns.
Documents
Cytiva CyDye Licensing Statements Cytiva General Licensing Statements
Additional Product Details
Specifikace
- Popis:PD-10 Spin adapter, for PD-10 desalting columns
- Sekvence:06