Spin Adapters for Gravity Columns
02, MiniSpin™ adapter, for PD MiniTrap™ G-25 columns
Katalogové číslo 28-9232-43
Dodavatel: Cytiva
Přejít na:
Product Details & Documents
Convenient and rapid cleanup with high reproducibility and the potential for high throughput by running multiple samples in parallel, High desalting capacity, The use of centrifugation protocol with all gravity columns.
- Convenient adapters, enable spinning of all gravity columns
Adapters are polypropylene rings designed for use with the gravity columns PD MiniTrap G-25, PD MidiTrap G-25, and PD-10 Desalting Columns when the spin protocol is used. MiniSpin Adapter fits PD MiniTrap G-25 columns, MidiSpin Adapter fits PD MidiTrap G-25 columns, and PD-10 Spin Adapter fits PD-10 Desalting Columns.
Documents
28923243_DATA FILE 28923244_DATA FILE 28923245_DATA FILE
Technický list
Cytiva CyDye Licensing Statements Cytiva General Licensing Statements
Další podrobnosti o produktu
Specifikace
- Popis:MiniSpin™ adapter, for PD MiniTrap™ G-25 columns
- Sekvence:02