Plugs for test tubes
10, 16 mm, Zelená
Katalogové číslo 212-0522
Dodavatel: BOETTGER PAUL
Přejít na:
Product Details & Documents
Polyethylene (PE). Ribbed plugs with two sealing ribs, very tight sealing.
- Not autoclavable
- Easy handling with a high, ribbed grip
- Easy to open by tilting 'one-hand operation'
Specifikace
- Barva:Zelená
- Sekvence:10
- Velikost:16 mm