Drum taps, PE
01, Fast outflow, Corrosive and aggressive chemicals, 3/4" Tri-Sure
Katalogové číslo MSGA2801
Dodavatel: asecos
Přejít na:
Product Details & Documents
These polyethylene taps are used for corrosive and aggressive chemicals.
- Fast outflow
- Tri-sure thread
Specifikace
- Závit:3/4" Tri-Sure
- Popis:Fast outflow
- Sekvence:01
- Určeno pro:Corrosive and aggressive chemicals