Orders LinkContactUsLinkComponent
Universal socks
Universal socks
Universal sorbent snakes, 15 per pack, 75×1200 mm
Katalogové číslo MSGA9954
Dodavatel:   asecos
Universal socks
Katalogové číslo MSGA9954
Dodavatel:  asecos
Universal socks
Universal socks
Universal sorbent snakes, 15 per pack, 75×1200 mm
Přejít na:
Price
2 350,00 Kč
Přihlásit se pro dostupnost
1 * 1 KS
Product Details & Documents
Universal socks for safe absorption of oil, water, cooling liquids and non aggressive chemicals.
  • Optimal for long term use
  • Capacity up to 16 times of its own weight
  • High rib and rub resistance
For containment and absorption of larger leaks.
Specifikace
  • Popis:
    Universal sorbent snakes
  • Balené:
    15 per pack
  • Ø×D:
    75×1200 mm