Orders LinkContactUsLinkComponent
Universal absorbent pillows
Universal absorbent pillows
Universal pillow, 10 pillows/pack, 460×460 mm
Katalogové číslo MSGA9943
Dodavatel:   asecos
Universal absorbent pillows
Katalogové číslo MSGA9943
Dodavatel:  asecos
Universal absorbent pillows
Universal absorbent pillows
Universal pillow, 10 pillows/pack, 460×460 mm
Přejít na:
Price
2 630,00 Kč
Přihlásit se pro dostupnost
1 * 1 KS
Product Details & Documents
Universal pillows for safe absorption of oil, water, cooling liquids and non aggressive chemicals.
  • Optimal for those hard to reach places
  • Capacity up to 16 times of its own weight
  • High rib and rub resistance
Specifikace
  • Popis:
    Universal pillow
  • D׊:
    460×460 mm
  • Balené:
    10 pillows/pack