Orders LinkContactUsLinkComponent
Microscope slides, Corning®
Microscope slides, Corning®
03, Frosted on both sides slides, Bezbarvá, Sklo, 75×25 mm
Katalogové číslo 734-4040
Dodavatel:   Corning
Microscope slides, Corning®
Katalogové číslo 734-4040
Dodavatel:  Corning
Microscope slides, Corning®
Microscope slides, Corning®
03, Frosted on both sides slides, Bezbarvá, Sklo, 75×25 mm
Přejít na:
Price
16 560,00 Kč
Přihlásit se pro dostupnost
1 * 1.440 KS
Product Details & Documents
These slides are either frosted on one side, on both sides, or plain.
  • Clarity reinforced water-white glass
  • Frosted slides are sandblasted, easy to write on and highly legible
  • Each slide is inspected for chips and rough edge before being shipped
Specifikace
  • Barva:
    Bezbarvá
  • Popis:
    Frosted on both sides slides
  • D׊:
    75×25 mm
  • Materiál:
    Sklo
  • Sekvence:
    03