Orders LinkContactUsLinkComponent
Microscope slides, Corning®
Microscope slides, Corning®
02, Frosted on one side slides, Bezbarvá, 02, Sklo, 75×25 mm
Catalog # 734-4039
Supplier:   Corning
Microscope slides, Corning®
Catalog # 734-4039
Supplier:  Corning
Microscope slides, Corning®
Microscope slides, Corning®
02, Frosted on one side slides, Bezbarvá, 02, Sklo, 75×25 mm
Price
12 710,00 Kč
Log in for availability
1 * 1.440 KS
Product Details & Documents
These slides are either frosted on one side, on both sides, or plain.
  • Clarity reinforced water-white glass
  • Frosted slides are sandblasted, easy to write on and highly legible
  • Each slide is inspected for chips and rough edge before being shipped
Specifications
  • Poč. jamek:
    02
  • Barva:
    Bezbarvá
  • Popis:
    Frosted on one side slides
  • D׊:
    75×25 mm
  • Materiál:
    Sklo
  • Sekvence:
    02