Orders LinkContactUsLinkComponent
Adapters for Huber circulators
Adapters for Huber circulators
14, Adapter, M24×1,5 female thread to M30×1,5 male thread
Catalog # HUBE6723
Supplier:   HUBER
Adapters for Huber circulators
Catalog # HUBE6723
Supplier:  HUBER
Adapters for Huber circulators
Adapters for Huber circulators
14, Adapter, M24×1,5 female thread to M30×1,5 male thread
Price
3 130,00 Kč
Log in for availability
1 * 1 KS
Product Details & Documents
Specifications
  • Popis:
    Adapter, M24×1,5 female thread to M30×1,5 male thread
  • Sekvence:
    14