Orders LinkContactUsLinkComponent
Adapters for Huber circulators
Adapters for Huber circulators
35, Adapter, M38×1,5 female thread to 3/4" male thread
Katalogové číslo HUBE6665
Dodavatel:   HUBER
Adapters for Huber circulators
Katalogové číslo HUBE6665
Dodavatel:  HUBER
Adapters for Huber circulators
Adapters for Huber circulators
35, Adapter, M38×1,5 female thread to 3/4" male thread
Přejít na:
Price
3 880,00 Kč
Přihlásit se pro dostupnost
1 * 1 KS
Product Details & Documents
Specifikace
  • Popis:
    Adapter, M38×1,5 female thread to 3/4" male thread
  • Sekvence:
    35