Orders LinkContactUsLinkComponent
Adapters for Huber circulators
Adapters for Huber circulators
32, Adapter, M30×1,5 female thread to 3/4" NPT male thread
Katalogové číslo HUBE6472
Dodavatel:   HUBER
Adapters for Huber circulators
Katalogové číslo HUBE6472
Dodavatel:  HUBER
Adapters for Huber circulators
Adapters for Huber circulators
32, Adapter, M30×1,5 female thread to 3/4" NPT male thread
Přejít na:
Price
2 880,00 Kč
Přihlásit se pro dostupnost
1 * 1 KS
Product Details & Documents
Specifikace
  • Popis:
    Adapter, M30×1,5 female thread to 3/4" NPT male thread
  • Sekvence:
    32