Adapters for Huber circulators
30, Adapter, M30×1,5 female thread to 3/4" female thread
Katalogové číslo HUBE6452
Dodavatel: HUBER
Přejít na:
Product Details & Documents
Specifikace
- Popis:Adapter, M30×1,5 female thread to 3/4" female thread
- Sekvence:30