Orders LinkContactUsLinkComponent
Accessories for compatible control refrigerated circulators
Accessories for compatible control refrigerated circulators
05, Displacement insert, W×D×H: 245×240×189 mm, CC-510, CC-515, CC-905, CC-905w, CC-906w
Katalogové číslo HUBE6050
Dodavatel:   HUBER
Accessories for compatible control refrigerated circulators
Katalogové číslo HUBE6050
Dodavatel:  HUBER
Accessories for compatible control refrigerated circulators
Accessories for compatible control refrigerated circulators
05, Displacement insert, W×D×H: 245×240×189 mm, CC-510, CC-515, CC-905, CC-905w, CC-906w
Přejít na:
Price
16 500,00 Kč
Přihlásit se pro dostupnost
1 * 1 KS
Product Details & Documents
Specifikace
  • Popis:
    Displacement insert, W×D×H: 245×240×189 mm
  • Sekvence:
    05
  • Určeno pro:
    CC-510, CC-515, CC-905, CC-905w, CC-906w