Orders LinkContactUsLinkComponent
Accessories for compatible control refrigerated circulators
Accessories for compatible control refrigerated circulators
04, Displacement insert, W×D×H: 265×145×185 mm, CC-510w, CC-515w, CC-520, CC-520w, CC-525w, CC-820, CC-820w
Katalogové číslo HUBE6049
Dodavatel:   HUBER
Accessories for compatible control refrigerated circulators
Katalogové číslo HUBE6049
Dodavatel:  HUBER
Accessories for compatible control refrigerated circulators
Accessories for compatible control refrigerated circulators
04, Displacement insert, W×D×H: 265×145×185 mm, CC-510w, CC-515w, CC-520, CC-520w, CC-525w, CC-820, CC-820w
Přejít na:
Price
14 750,00 Kč
Přihlásit se pro dostupnost
1 * 1 KS
Product Details & Documents
Specifikace
  • Popis:
    Displacement insert, W×D×H: 265×145×185 mm
  • Sekvence:
    04
  • Určeno pro:
    CC-510w, CC-515w, CC-520, CC-520w, CC-525w, CC-820, CC-820w