FLUID CALIBRATION 2000 AW 2000 MPAS 20C 1 * 100 mL
Katalogové číslo HAAK082-5304
Dodavatel: HAAKE
Frequently Bought Together
Detergents, liquid, for automated cleaning of glassware, neodisher® FLA
Dodavatel: Dr. Weigert
Aceton ≥99%, TECHNICAL
Dodavatel: VWR Chemicals
Voda, LiChrosolv® for chromatography, for LC-MS, Supelco®
Dodavatel: Merck
VWR®, Nitrile Gloves
Dodavatel: VWR Collection
Acetonitril ≥99.9%, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Dodavatel: VWR Chemicals
[EN]SYRINGE 20ML SLIP TIP ECCENTRIC 1 * 50 KS
Dodavatel: TERUMO
Aceton ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Dodavatel: VWR Chemicals
Chlorid sodný, EMSURE® ACS, ISO, Reag. Ph. Eur. p.a., Supelco®
Dodavatel: Merck
Neutralising agent for the automated cleaning of glassware, neodisher® Z
Dodavatel: Dr. Weigert
Acetonitril ≥99.9%, HiPerSolv CHROMANORM®, gradient grade for HPLC
Dodavatel: VWR Chemicals
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Dodavatel: VWR Chemicals
Voda, HiPerSolv CHROMANORM® for HPLC
Dodavatel: VWR Chemicals
Voda, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Dodavatel: VWR Chemicals