Stoppers, flange plug caps
Žlutá, 16 mm
Katalogové číslo SIMPT404-10Y
Dodavatel: Simport Scientific
Přejít na:
Product Details & Documents
PE Plug type push-in caps and 2-position snap caps are made of polyethylene for test tubes with outside diameters of 12 or 17 mm. Dual position caps offer two possibilities: the closed but unsealed position where samples are maintained aerobic for microbiological procedures; and the fully sealed position where the cap is pushed tight to seal the tube for anaerobic use or for storage, transfer and centrifuge applications.
Specifikace
- Ø dno:16 mm
- Barva:Žlutá