ZIRCONIUM ACETATE, SOLUTION IN DILUTE AC 1 * 500 mL
Catalog # SIAL413801-500ML
Supplier: Merck
Frequently Bought Together
1-Butanol ≥98.5%, GPR RECTAPUR®
Supplier: VWR Chemicals
Dichlormethan ≥98% stabilised, TECHNICAL
Supplier: VWR Chemicals
Cyklohexan, EMSURE® ACS, ISO, Reag. Ph. Eur. p.a., Supelco®
Supplier: Merck
n-Heptan, EMPLURA®, Supelco®
Supplier: Merck
Glycerin ≥99.5%, AnalaR NORMAPUR® ACS analytical reagent, redistilled
Supplier: VWR Chemicals
AMMONIUM ACETATE, GR ACS 1 * 500 g
Supplier: Merck
(±)-2-Butanol, EMSURE® p.a., Supelco®
Supplier: Merck
Izopropylacetát for synthesis, Sigma-Aldrich®
Supplier: Merck
Acetonitril, anhydrous (max. 0.001% H₂O)
Supplier: Merck
Dichlormethan, anhydrous (max. 0.001% H₂O) ≥99.8% stabilised
Supplier: VWR Chemicals
Glycerin 87% 86%, AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
Petrolether, 80…110 °C, TECHNICAL
Supplier: VWR Chemicals
N,N-Dimethylformamid ≥99.5%, GPR RECTAPUR®
Supplier: VWR Chemicals
n-Heptan, extra dry over Molecular Sieve 99+%, AcroSeal®
Supplier: Thermo Fisher Scientific