Orders LinkContactUsLinkComponent
Accessories for automatic burettes, Pellet pattern
Accessories for automatic burettes, Pellet pattern
01, Reservoir bottle, glass, clear, 2000 ml, with 29/32 joint
Katalogové číslo BIBB3297/12
Dodavatel:   SciLabware
Accessories for automatic burettes, Pellet pattern
Katalogové číslo BIBB3297/12
Dodavatel:  SciLabware
Accessories for automatic burettes, Pellet pattern
Accessories for automatic burettes, Pellet pattern
01, Reservoir bottle, glass, clear, 2000 ml, with 29/32 joint
Přejít na:
Price
1 740,00 Kč
Přihlásit se pro dostupnost
1 * 1 KS
Product Details & Documents
Specifikace
  • Popis:
    Reservoir bottle, glass, clear, 2000 ml, with 29/32 joint
  • Sekvence:
    01