Orders LinkContactUsLinkComponent
Cold trap, type S
Cold trap, type S
02, Cold trap, S 29-GL-Z, borosilicate glass 3,3, with threaded glass joint GL 18, 150 ml condensate capacity and length of 200 mm
Katalogové číslo KGWA17407
Dodavatel:   KGW
Cold trap, type S
Katalogové číslo KGWA17407
Dodavatel:  KGW
Cold trap, type S
02, Cold trap, S 29-GL-Z, borosilicate glass 3,3, with threaded glass joint GL 18, 150 ml condensate capacity and length of 200 mm
Přejít na:
Price
11 520,00 Kč
Přihlásit se pro dostupnost
1 * 1 KS
Product Details & Documents
Type S cold trap has a conical ground joint NS 45/40 as connection part. The condensation area can be separated from the upper section of cold trap. Therefore it is easy to pour out the condensate and to clean the whole cold trap. The connection part NS 45/40 is secured by a spring clamp out of steel.
Certifications: Made from borosilicate glass 3.3, in compliance with DIN/ISO 3585.
Specifikace
  • Popis:
    Cold trap, S 29-GL-Z, borosilicate glass 3,3, with threaded glass joint GL 18, 150 ml condensate capacity and length of 200 mm
  • Sekvence:
    02