Orders LinkContactUsLinkComponent
Posts for shelving systems, solid, Super Erecta® SiteSelect™
Posts for shelving systems, solid, Super Erecta® SiteSelect™
15, Stationary post, SiteSelect™, metroseal 3, 1590 mm height, Super Adjustable Super Erecta® shelving, Super Erecta® shelving, Super Erecta Pro™ shelving
Katalogové číslo METO63PK3
Dodavatel:   Metro International
Posts for shelving systems, solid, Super Erecta® SiteSelect™
Katalogové číslo METO63PK3
Dodavatel:  Metro International
Posts for shelving systems, solid, Super Erecta® SiteSelect™
Posts for shelving systems, solid, Super Erecta® SiteSelect™
15, Stationary post, SiteSelect™, metroseal 3, 1590 mm height, Super Adjustable Super Erecta® shelving, Super Erecta® shelving, Super Erecta Pro™ shelving
Přejít na:
Price
1 280,00 Kč
Přihlásit se pro dostupnost
1 * 1 KS
Product Details & Documents
Specifikace
  • Popis:
    Stationary post, SiteSelect™, metroseal 3, 1590 mm height
  • Sekvence:
    15
  • Určeno pro:
    Super Adjustable Super Erecta® shelving, Super Erecta® shelving, Super Erecta Pro™ shelving